Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERMAP Antibody (767505), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347510 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347510 Supplier Novus Biologicals Supplier No. FAB49281MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERMAP Monoclonal specifically detects ERMAP in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen ERMAP
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 767505
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias erythroblast membrane-associated protein, erythroblast membrane-associated protein (RD and SC blood groups), erythroblast membrane-associated protein (Scianna blood group), erythroid membrane-associated protein, hERMAP, MGC118810, MGC118811, PRO2801, Radin blood group, Radin blood group (Rd), Radin blood group antigen, RDMGC118812, Scianna blood group, Scianna blood group (Sc), Scianna blood group antigen, SCMGC118813
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ERMAP, His30-Ala155, Accession # Q96PL5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114625
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.