Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERMAP Antibody (OTIC8), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N272339P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272339P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272339P594 Supplier Novus Biologicals Supplier No. NBP272339PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERMAP Monoclonal specifically detects ERMAP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ERMAP
Applications Flow Cytometry
Classification Monoclonal
Clone OTIC8
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias erythroblast membrane-associated protein, erythroblast membrane-associated protein (RD and SC blood groups), erythroblast membrane-associated protein (Scianna blood group), erythroid membrane-associated protein, hERMAP, MGC118810, MGC118811, PRO2801, Radin blood group, Radin blood group (Rd), Radin blood group antigen, RDMGC118812, Scianna blood group, Scianna blood group (Sc), Scianna blood group antigen, SCMGC118813
Gene Symbols ERMAP
Host Species Mouse
Immunogen Full length recombinant protein of human ERMAP (NP_061008) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 114625.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.