Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERMN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156953
Description
ERMN Polyclonal specifically detects ERMN in Human samples. It is validated for Western Blot.Specifications
| ERMN | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ERMIN, ermin, ERM-like protein, JNKIAA1189ermin, juxtanodin | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57471 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8TAM6-2 | |
| ERMN | |
| Synthetic peptides corresponding to KIAA1189 The peptide sequence was selected from the middle region of KIAA1189. Peptide sequence RVIEFKKKHEEVSQFKEEGDASEDSPLSSASSQAVTPDEQPTLGKKSDIS. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Sumatran orangutan: 100%; Human: 100%;. | |
| Human, Rat, Canine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction