Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERMN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ERMN |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ERMN Polyclonal specifically detects ERMN in Human samples. It is validated for Western Blot.Specifications
| ERMN | |
| Polyclonal | |
| Rabbit | |
| Q8TAM6-2 | |
| 57471 | |
| Synthetic peptides corresponding to KIAA1189 The peptide sequence was selected from the middle region of KIAA1189. Peptide sequence RVIEFKKKHEEVSQFKEEGDASEDSPLSSASSQAVTPDEQPTLGKKSDIS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ERMIN, ermin, ERM-like protein, JNKIAA1189ermin, juxtanodin | |
| ERMN | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title