Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERO1L Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB003467 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003467 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003467 Supplier Novus Biologicals Supplier No. NB1002525V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERO1L Polyclonal specifically detects ERO1L in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen ERO1L
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein, ERO1 (S. cerevisiae)-like, ERO1A, ERO1-alpha, ERO1-L, ERO1-L-alpha, ERO1-like (S. cerevisiae), ERO1-like protein alpha, oxidoreductin-1-L-alpha
Gene Symbols ERO1A
Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal region of human ERO1L [UniProt Q96HE7]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, ER Markers, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.