Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERO1L Antibody (V23P3C9*H2), HRP, Novus Biologicals™
SDP

Catalog No. NBP250388H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250388H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250388H Supplier Novus Biologicals Supplier No. NBP250388H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERO1L Monoclonal specifically detects ERO1L in Human, Mouse, Rat, Porcine, Canine, Equine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen ERO1L
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V23P3C9*H2
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein, ERO1 (S. cerevisiae)-like, ERO1A, ERO1-alpha, ERO1-L, ERO1-L-alpha, ERO1-like (S. cerevisiae), ERO1-like protein alpha, oxidoreductin-1-L-alpha
Gene Symbols ERO1A
Host Species Mouse
Immunogen Peptide Sequence - KLIANMPESG (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), sheep (Ovis aries), bovine (Bos taurus), canine (Canis familiaris), porcine (Sus scrofa) and equine (Equus caballus)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, ER Markers, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 30001
Target Species Human, Mouse, Rat, Pig, Canine, Equine, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.