Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERP29 Antibody, Novus Biologicals™
SDP

Catalog No. NBP105028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105028 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105028 Supplier Novus Biologicals Supplier No. NBP105028
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ERP29 Polyclonal specifically detects ERP29 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen ERP29
Applications Western Blot, Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-0.3ug/ml, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006808.1
Gene Alias C12orf8chromosome 12 open reading frame 8, endoplasmic reticulum lumenal protein ERp28, endoplasmic reticulum protein 29, Endoplasmic reticulum resident protein 28, endoplasmic reticulum resident protein 29, ERp28Endoplasmic reticulum resident protein 31, ERp29, ERp31ERP28, PDIA9, PDI-DB, protein disulfide isomerase family A, member 9
Gene Symbols ERP29
Host Species Goat
Immunogen Peptide with sequence C-EKNKMSDGKKEELQ corresponding to C-Terminus according to NP_006808.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10961
Test Specificity This antibody is expected to recognize isoform 1 (NP_006808.1) only.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.