Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERP44 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB080597 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080597 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080597 Supplier Novus Biologicals Supplier No. NBP297000F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERP44 Polyclonal specifically detects ERP44 in Human samples. It is validated for ELISA.

Specifications

Antigen ERP44
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias endoplasmic reticulum protein 44, endoplasmic reticulum resident protein 44 kDa, ER protein 44, ERp44, KIAA0573endoplasmic reticulum resident protein 44, PDIA10, protein disulfide isomerase family A, member 10, thioredoxin domain containing 4 (endoplasmic reticulum), Thioredoxin domain-containing protein 4, TXNDC4
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ERP44 (NP_055866.1; Met1-Asp402).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23071
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.