Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERp57/PDIA3 Antibody (OTI3E1), Biotin, Novus Biologicals™
SDP

Catalog No. NBP270848B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270848B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270848B Supplier Novus Biologicals Supplier No. NBP270848B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERp57/PDIA3 Monoclonal specifically detects ERp57/PDIA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ERp57/PDIA3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3E1
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3
Gene Symbols PDIA3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids PDIA3(NP_005304) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cellular Markers, ER Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 2923.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.