Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERp57/PDIA3 Mouse anti-Bovine, Canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Porcine, Rabbit, Rat, Clone: Map.ERp57 (GRP58), Novus Biologicals™
SDP

Catalog No. NB5968950UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB5968950UG 50 μg
NB59689200U 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB5968950UG Supplier Novus Biologicals Supplier No. NBP25968950UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERp57/PDIA3 Monoclonal specifically detects ERp57/PDIA3 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen ERp57/PDIA3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Map.ERp57 (GRP58)
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100
Formulation PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3
Gene Symbols PDIA3
Host Species Mouse
Immunogen Human recombinant ERp57 (Grp58)
Molecular Weight of Antigen 57 kDa
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cellular Markers, ER Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 2923
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig, Hamster, Monkey, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.