Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERp72 Antibody (004), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB038094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038094 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038094 Supplier Novus Biologicals Supplier No. NBP290207AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ERp72 Monoclonal specifically detects ERp72 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ERp72
Applications Immunofluorescence
Classification Monoclonal
Clone 4
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry/Immunofluorescence, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Endoplasmic reticulum resident protein 70, Endoplasmic reticulum resident protein 72, ER protein 70, ERp70, ERP70ER protein 72, ERp72, ERp-72, ERP72EC 5.3.4.1, intestinal-related), protein disulfide isomerase family A, member 4, protein disulfide isomerase related protein (calcium-binding protein, protein disulfide isomerase-associated 4, protein disulfide-isomerase A4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ERp72 (P13667; Met1-Thr641).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9601
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.