Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERp72 Antibody (OTI2B9), PerCP, Novus Biologicals™
SDP

Catalog No. NB288840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288840 Supplier Novus Biologicals Supplier No. NBP271488PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERp72 Monoclonal antibody specifically detects ERp72 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen ERp72
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B9
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Endoplasmic reticulum resident protein 70, Endoplasmic reticulum resident protein 72, ER protein 70, ERp70, ERP70ER protein 72, ERp72, ERp-72, ERP72EC 5.3.4.1, intestinal-related), protein disulfide isomerase family A, member 4, protein disulfide isomerase related protein (calcium-binding protein, protein disulfide isomerase-associated 4, protein disulfide-isomerase A4
Host Species Mouse
Immunogen Full length human recombinant protei of human PDIA4(NP_004902) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9601
Target Species Human, Mouse, Rat, Canine, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.