Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErpN/OspE Antibody, Novus Biologicals™
SDP

Catalog No. NBP221682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221682 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221682 Supplier Novus Biologicals Supplier No. NBP221682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ErpN/OspE Polyclonal specifically detects ErpN/OspE in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ErpN/OspE
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:5000
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. H7C7N5
Gene Alias erpA8, BB_L39, ErpA8 protein
Gene Symbols erpN
Host Species Rabbit
Immunogen MBP-fusion protein corresponding to Borrelia burgdorferi ErpN/OspE protein. (Uniprot: H7C7N5)
Molecular Weight of Antigen 19.57 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1194664
Test Specificity It is directed against, and shows specific reactivity for, Borrelia burgdorferi OspE protein.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Bacteria
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.