Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERR alpha Mouse anti-Human, Unconjugated, Clone: 2ERR10, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | ERR alpha |
---|---|
Clone | 2ERR10 |
Applications | ChIP Assay, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE18912MI
|
MilliporeSigma
MABE189125UG |
25 μg |
Each for $182.31
|
|
|||||
MABE18911MI
|
MilliporeSigma
MABE1891100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
ERR alpha Monoclonal antibody specifically detects ERR alpha in Human samples. It is validated for Chromatin Immunoprecipitation (ChIP), ELISA, Immunocytochemistry, Immunoprecipitation, Western Blotting.Specifications
ERR alpha | |
ChIP Assay, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Steroid hormone receptor ERR1;Estrogen receptor-like 1;Estrogen-related receptor alpha;Nuclear receptor subfamily 3 group B member 1 | |
His-tagged full length human recombinant Steroid hormone receptor ERR alpha (ERR1). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼53 kDa observed; 45.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
2ERR10 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_001269379 | |
ESRRA, ERR1, ESRL1, NR3B1 | |
IgG1 κ | |
Protein G Purified | |
Clone 2ERR10 is a mouse monoclonal antibody that detects human Steroid hormone receptor ERR alpha (ERR1). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title