Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERR alpha/NR3B1 Antibody (6F1E6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61768 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61768 Supplier Novus Biologicals Supplier No. NBP261768
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERR alpha/NR3B1 Monoclonal specifically detects ERR alpha/NR3B1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen ERR alpha/NR3B1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 6F1E6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias ERR1Estrogen receptor-like 1, ERRa, ESRL1ERR-alpha, estrogen-related receptor alphaERRalpha, NR3B1Nuclear receptor subfamily 3 group B member 1, steroid hormone receptor ERR1
Gene Symbols ESRRA
Host Species Mouse
Immunogen Purified recombinant fragment of human ESRRA (AA: 198-376) expressed in E. Coli.
Molecular Weight of Antigen 66.2 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2101
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.