Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERR alpha/NR3B1 Antibody (6F1E6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | ERR alpha/NR3B1 |
---|---|
Clone | 6F1E6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61768
![]() |
Novus Biologicals
NBP261768 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
ERR alpha/NR3B1 Monoclonal specifically detects ERR alpha/NR3B1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
ERR alpha/NR3B1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Human, Rat | |
2101 | |
Purified recombinant fragment of human ESRRA (AA: 198-376) expressed in E. Coli. | |
Primary | |
66.2 kDa |
6F1E6 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
Monoclonal | |
Purified | |
Cancer, Cell Biology, Cell Cycle and Replication, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
ERR1Estrogen receptor-like 1, ERRa, ESRL1ERR-alpha, estrogen-related receptor alphaERRalpha, NR3B1Nuclear receptor subfamily 3 group B member 1, steroid hormone receptor ERR1 | |
ESRRA | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title