Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERR alpha/NR3B1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB338371 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338371 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338371 Supplier Novus Biologicals Supplier No. NBP147254MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERR alpha/NR3B1 Polyclonal antibody specifically detects ERR alpha/NR3B1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ERR alpha/NR3B1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ERR1Estrogen receptor-like 1, ERRa, ESRL1ERR-alpha, estrogen-related receptor alphaERRalpha, NR3B1Nuclear receptor subfamily 3 group B member 1, steroid hormone receptor ERR1
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human Estrogen Related Receptor alpha (within residues 1-50). [Swiss-Prot# P11474]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2101
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.