Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERR beta/NR3B2 Antibody, Novus Biologicals™
SDP

Catalog No. NB300748 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300748 Supplier Novus Biologicals Supplier No. NB300748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERR beta/NR3B2 Polyclonal specifically detects ERR beta/NR3B2 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ERR beta/NR3B2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS with 1mg/ml BSA with 0.05% Sodium Azide
Gene Alias estrogen-related receptor beta
Gene Symbols ESRRB
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues D(376) Y E L S Q R H E E P R R(388) of mouse ERR2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2103
Test Specificity Estrogen-Related Receptor beta
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.