Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERRFI1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096158 20 μL
NB096157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096158 Supplier Novus Biologicals Supplier No. NBP30487020UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

ERRFI1 Polyclonal specifically detects ERRFI1 in Rat samples. It is validated for Western Blot.

Specifications

Antigen ERRFI1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ERBB receptor feedback inhibitor 1, GENE-33, MIG6, MIG-6Mitogen-inducible gene 6 protein, RALTreceptor-associated late transducer
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 223-462 of human ERRFI1 (NP_061821.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54206
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.