Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythrocyte Specific Antibody (SFL23.6), Biotin, Novus Biologicals™
SDP

Catalog No. NBP234715B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234715B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234715B Supplier Novus Biologicals Supplier No. NBP234715B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Erythrocyte Specific Monoclonal specifically detects Erythrocyte Specific in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Erythrocyte Specific
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone SFL23.6
Conjugate Biotin
Dilution Immunocytochemistry/Immunofluorescence
Gene Alias erythroid cell surface antigen
Host Species Mouse
Immunogen Hepatocytes from 20-22 week-old human fetus
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Mouse Anti-Human Erythrocytes Clone SFL23.6 monoclonal antibody is directed against an erythroid cell surface antigen. It shows a well-defined reactivity with cells of the erythroid lineage at all stages of maturation in the peripheral blood, bone marrow, and fetal liver. Non-erythroid lineages are negative by flow cytometry. Although the exact identity of the antigen has yet to be determined, it has been shown to be distinct from Glycophorin A. This monoclonal antibody is useful in the diagnosis of erythroleukemia, identification of bone marrow erythroid precursors, gating erythroid nucleated precursor cells from malignant cells in bone marrow specimens.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.