Learn More
Description
Specifications
Specifications
| Antigen | Erythrocyte Specific |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SFL23.6 |
| Conjugate | PE-Cyanine7 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | erythroid cell surface antigen |
| Host Species | Mouse |
| Immunogen | Hepatocytes from 20-22 week-old human fetus |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
