Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin/EPO Antibody (10F3), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP221994G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221994G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221994G Supplier Novus Biologicals Supplier No. NBP221994G

Rat Monoclonal Antibody

Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Mouse samples. It is validated for ELISA.

Specifications

Antigen Erythropoietin/EPO
Applications ELISA
Classification Monoclonal
Clone 10F3
Conjugate DyLight 488
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Gene Symbols EPO
Host Species Rat
Immunogen Mouse recombinant Erythropoietin/EPO
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 2056
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.