Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin/EPO Antibody (EPO/1368) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB254537100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254537100 100 μg
NB254537200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254537100 Supplier Novus Biologicals Supplier No. NBP254537100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.

Specifications

Antigen Erythropoietin
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone EPO/1368
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 4 - 8 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias EP, epoetin, erythropoietin, MGC138142, MVCD2
Gene Symbols EPO
Host Species Mouse
Immunogen Recombinant fragment of human EPO protein (aa28-162) (exact sequence is proprietary)
Molecular Weight of Antigen 37 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 2056
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.