Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin R Antibody (016), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB108508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108508 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108508 Supplier Novus Biologicals Supplier No. NBP289615AF488

Rabbit Monoclonal Antibody

Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Western Blot.

Specifications

Antigen Erythropoietin R
Applications Western Blot
Classification Monoclonal
Clone 16
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias EPO-R, erythropoietin receptor, MGC138358
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Erythropoietin R (NP_000112.1; Met1-Pro250).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2057
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.