Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin R Antibody (38409), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348544 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348544 Supplier Novus Biologicals Supplier No. FAB307MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Erythropoietin R Monoclonal antibody specifically detects Erythropoietin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation

Specifications

Antigen Erythropoietin R
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 38409
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias EPO-R, erythropoietin receptor, MGC138358
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R, Pro26-Pro250, Accession # P19235
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2057
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.