Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin R Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP119388A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP119388A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP119388A32 Supplier Novus Biologicals Supplier No. NBP119388AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Erythropoietin R Polyclonal specifically detects Erythropoietin R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Erythropoietin R
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EPO-R, erythropoietin receptor, MGC138358
Gene Symbols EPOR
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human EPO receptor protein (between residues 300-400) [UniProt P19235]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2057
Test Specificity EpoR (A364) detects endogenous levels of EpoR protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.