Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin R Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP119388SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119388SS 0.025 mL
NBP119388 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP119388SS Supplier Novus Biologicals Supplier No. NBP119388SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Erythropoietin R Polyclonal specifically detects Erythropoietin R in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Erythropoietin R
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.0 ug/ml, Simple Western 1:10, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P19235
Gene Alias EPO-R, erythropoietin receptor, MGC138358
Gene Symbols EPOR
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human EPO receptor protein (between residues 300-400) [UniProt P19235]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2057
Test Specificity EpoR (A364) detects endogenous levels of EpoR protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.