Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erythropoietin R Rabbit anti-Mouse, Clone: 140, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290367100UL
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Mouse samples. It is validated for ELISA.Specifications
Erythropoietin R | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
140 | |
ELISA 1:5000-1:10000 | |
EPO-R, erythropoietin receptor, MGC138358 | |
Recombinant Mouse Erythropoietin R Protein | |
100 μg | |
Angiogenesis, Cancer, Immunology, Phospho Specific, Signal Transduction | |
2057 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction