Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erythropoietin R Rabbit anti-Mouse, Clone: 140, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | Erythropoietin R |
---|---|
Clone | 140 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Mouse samples. It is validated for ELISA.Specifications
Erythropoietin R | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
EPO-R, erythropoietin receptor, MGC138358 | |
Recombinant Mouse Erythropoietin R Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
140 | |
ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Immunology, Phospho Specific, Signal Transduction | |
0.2 um filtered solution in PBS | |
2057 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title