Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythrosin Yellowish, Thermo Scientific™

Catalog No. AAA1562614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1562614 25 g
AAA1562622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1562614 Supplier Thermo Scientific Chemicals Supplier No. A1562614

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33239-19-9
Molecular Formula C20H8I2Na2O5
Molecular Weight (g/mol) 628.07
MDL Number MFCD00036461
InChI Key AHSJNHONMVUMLK-UHFFFAOYSA-L
Synonym C.I. 45425; Acid red 95
IUPAC Name disodium 4',5'-diiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
SMILES [Na+].[Na+].[O-]C1=CC=C2C(OC3=C(I)C([O-])=CC=C3C22OC(=O)C3=CC=CC=C23)=C1I

Specifications

Melting Point >300°C
Formula Weight 628.07
Sensitivity Light sensitive
Quantity 25 g
Chemical Name or Material Erythrosin Yellowish
EINECSNumber 251-419-2
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.