Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUJIFILM Wako PYROSTAR™ ES-F 2 mL Bulk Kit

Catalog No. 12333017 Shop All FujiFilm Products
Click to view available options
Sensitivity:
0.015 EU/mL
0.03 EU/mL
0.06 EU/mL
0.125 EU/mL
0.25 EU/mL

Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, Dual-purpose formulated to be used as gel clot or kinetic turbidimetric assay, Bulk configuration for large volume labs

  • Endotoxin-specific reagent avoids false positive results from glucans
  • Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
  • Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
  • Due to high sensitivity, chances of interference are reduced
  • Facilitates easier transition from gel clot to KTA
TRUSTED_SUSTAINABILITY

Specifications

Form Lyophilized
Fill Volume 2 mL
Includes 100 x 2 mL vials of lysate
Quantity 100 Vials
Sensitivity 0.125 EU/mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.