Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUJIFILM Wako PYROSTAR™ ES-F Single Test
Click to view available options
Sensitivity:
0.015 EU/mL
0.03 EU/mL
0.125 EU/mL
0.25 EU/mL
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- KTA sensitivities range from 0.001 to 10 EU/mL
- Can be used with the traditional water bath or heat block
- Can be used with the Toxinometer™ to facilitate easier transition from gel clot to KTA
Specifications
Specifications
| Form | Lyophilized |
| Fill Volume | 200 μL |
| Includes | 25 single test vials and 1 vial of CSE (500 ng/vial) |
| Quantity | 25 Tests |
| Sensitivity | 0.03 EU/mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction