Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUJIFILM Wako PYROSTAR™ ES-F Single Test

Catalog No. 12333013
Encompass
Click to view available options
Sensitivity:
0.015 EU/mL
0.03 EU/mL
0.125 EU/mL
0.25 EU/mL

Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, Includes pre-dispensed LAL reagent for single measurement, for use with Toxinometer™ and to assist labs transitioning from gel clot to KTA methodology

  • Endotoxin-specific reagent avoids false positive results from glucans
  • KTA sensitivities range from 0.001 to 10 EU/mL
  • Can be used with the traditional water bath or heat block
  • Can be used with the Toxinometer™ to facilitate easier transition from gel clot to KTA

Specifications

Form Lyophilized
Fill Volume 200 μL
Includes 25 single test vials and 1 vial of CSE (500 ng/vial)
Quantity 25 Tests
Sensitivity 0.25 EU/mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.