Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESAM Antibody (1021723), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB370710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370710 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370710 Supplier Novus Biologicals Supplier No. FAB42042X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ESAM Monoclonal specifically detects ESAM in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen ESAM
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 1021723
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ESAM, Gln30-Ala247, Accession # Q96AP7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90952
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.