Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESAM Antibody (110), Alexa Fluor™ 488
SDP

Catalog No. NB077444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB077444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB077444 Supplier Novus Biologicals Supplier No. NBP290757AF488

Rabbit Monoclonal Antibody

ESAM Monoclonal specifically detects ESAM in Mouse samples. It is validated for ELISA.

Specifications

Antigen ESAM
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ESAM (Q925F2; Met1-Ala251).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90952
Test Specificity No cross-reactivity in ELISA with: Human ESAM
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.