Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRP1 Antibody (27H12.F3.H5), Novus Biologicals™
SDP

Catalog No. NBP142778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142778 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142778 Supplier Novus Biologicals Supplier No. NBP142778
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ESRP1 Monoclonal specifically detects ESRP1 in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ESRP1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 27H12
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:750000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q3US41
Gene Alias epithelial splicing regulatory protein 1, FLJ20171, RBM35A, RMB35A, RNA binding motif protein 35A, RNA-binding motif protein 35A, RNA-binding protein 35A
Gene Symbols ESRP1
Host Species Mouse
Immunogen ESRP1 Antibody (27H12.F3.H5) was produced by repeated immunizations of full length recombinant mouse ESRP1 fusion protein. (Uniprot: Q3US41)
Molecular Weight of Antigen 75.55 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54845
Test Specificity This antibody reacts with mouse ESRP1 protein. A BLAST analysis of the immunizing protein sequence shows 100% homology with ESRP1 from mouse and a 91% sequence homology with ESRP1 from human, pig, rat, opossum, horse, cattle, panda, dog, and chimpanzee. The binding epitope of this monoclonal antibody has not been mapped.
Target Species Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.