Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRP2 Antibody (22C5.H7.A6), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB151249 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151249 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151249 Supplier Novus Biologicals Supplier No. NBP177972AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ESRP2 Monoclonal specifically detects ESRP2 in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ESRP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 22C5.H7.A6
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias epithelial splicing regulatory protein 2, FLJ21918, FLJ22248, RBM35BRNA-binding protein 35B, RNA binding motif protein 35A, RNA binding motif protein 35B, RNA-binding motif protein 35B
Host Species Mouse
Immunogen ESRP2 Antibody (22C5.H7.A6) is produced by repeated immunizations of full length recombinant mouse Esrp-2 fusion protein. (Uniprot: Q8K0G8)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80004
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.