Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESX1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB356569 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356569 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356569 Supplier Novus Biologicals Supplier No. NBP177114MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ESX1 Polyclonal antibody specifically detects ESX1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ESX1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ESX homeobox 1, ESX1LESX1R, ESX1-related protein, ESXR1homeobox protein ESX1, Extraembryonic, spermatogenesis, homeobox 1, extraembryonic, spermatogenesis, homeobox 1 homolog, extraembryonic, spermatogenesis, homeobox 1 homolog (mouse)
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the center of human ESX1. The immunogen is located within amino acids 140 - 190 of ESX1. Amino Acid Squence: QFQLQELENFFDESQYPDV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80712
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.