Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | ESX1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18021420
![]() |
Novus Biologicals
NBP18021420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180214
![]() |
Novus Biologicals
NBP180214 |
100 μL |
Each for $499.50
|
|
|||||
Description
ESX1 Polyclonal specifically detects ESX1 in Human samples. It is validated for Western Blot.Specifications
ESX1 | |
Polyclonal | |
Purified | |
RUO | |
NP_031983 | |
80712 | |
Synthetic peptide directed towards the N terminal of mouse ESX1. Peptide sequence MESHKKCPCCYCTDLKTFVGAVKEETLQDPQPSLSSTLLEGADYQENAES. | |
Primary | |
35 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
ESX homeobox 1, ESX1LESX1R, ESX1-related protein, ESXR1homeobox protein ESX1, Extraembryonic, spermatogenesis, homeobox 1, extraembryonic, spermatogenesis, homeobox 1 homolog, extraembryonic, spermatogenesis, homeobox 1 homolog (mouse) | |
ESX1 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title