Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETFA Antibody (OTI5C4), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270693V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270693V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270693V Supplier Novus Biologicals Supplier No. NBP270693V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ETFA Monoclonal specifically detects ETFA in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ETFA
Applications Western Blot
Classification Monoclonal
Clone OTI5C4
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias alpha-ETF, electron transfer flavoprotein alpha-subunit, Electron transfer flavoprotein subunit alpha, mitochondrial, electron-transfer-flavoprotein, alpha polypeptide, EMA, GA2, glutaric aciduria II, MADD
Gene Symbols ETFA
Host Species Mouse
Immunogen Full length recombinant protein of human ETFA (NP_000117) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2108.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.