Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETFA Antibody (OTI5C4), FITC, Novus Biologicals™
SDP

Catalog No. NB289443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289443 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289443 Supplier Novus Biologicals Supplier No. NBP270693F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ETFA Monoclonal antibody specifically detects ETFA in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ETFA
Applications Western Blot
Classification Monoclonal
Clone OTI5C4
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias alpha-ETF, electron transfer flavoprotein alpha-subunit, Electron transfer flavoprotein subunit alpha, mitochondrial, electron-transfer-flavoprotein, alpha polypeptide, EMA, GA2, glutaric aciduria II, MADD
Host Species Mouse
Immunogen Full length human recombinant protein of human ETFA (NP_000117) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2108
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.