Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETFDH Antibody, Novus Biologicals™
SDP

Catalog No. NBP152059 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152059 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152059 Supplier Novus Biologicals Supplier No. NBP152059
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ETFDH Polyclonal specifically detects ETFDH in Human, Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen ETFDH
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_004444.2
Gene Alias electron-transferring-flavoprotein dehydrogenaseEC 1.5.5.1, ETFQO, ETF-QO, MADD, mitochondrial
Gene Symbols ETFDH
Host Species Goat
Immunogen Peptide with sequence C-EHDQPAHLTLRD corresponding to internal region according to NP_004444.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2110
Test Specificity Cross reacts with Cross reacts with Human, Cross reacts with Mouse and Rat. Expected from sequence similarity: Human, Mouse, Rat, Dog, Cow and Rabbit.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.