Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanesulfonic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
594-45-6
C2H6O3S
110.13
MFCD00007529
CCIVGXIOQKPBKL-UHFFFAOYSA-N
ethylsulfonic acid,ethanesulphonic acid,ethane sulfonate,ethane sulfonic acid,ethane sulphonic acid,ethansulfonic acid,ethylsulphonic acid,1-ethanesulfonic acid,2-ethanesulfonic acid,ethanesulfonicacid
11668
CHEBI:42465
ethanesulfonic acid
CCS(=O)(=O)O
594-45-6
C2H6O3S
110.13
MFCD00007529
CCIVGXIOQKPBKL-UHFFFAOYSA-N
ethylsulfonic acid,ethanesulphonic acid,ethane sulfonate,ethane sulfonic acid,ethane sulphonic acid,ethansulfonic acid,ethylsulphonic acid,1-ethanesulfonic acid,2-ethanesulfonic acid,ethanesulfonicacid
11668
CHEBI:42465
ethanesulfonic acid
CCS(=O)(=O)O
Boiling Point 136°C to 140°C (3.0 hPa)
Flash Point >110°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow to Brown
Melting Point -17.0°C
Specific Gravity 1.343
Quantity 100g
Refractive Index 1.4325 to 1.4355
Density 1.3430g/mL
Assay Percent Range 95%
Linear Formula C2H5SO3H
Beilstein 04, 5
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 110.13
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Ethanesulfonic acid, 95%
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger