Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanesulfonic acid, 95%

Catalog No. p-2691415
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 594-45-6 | C2H5O3S | 109.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-45-6
Molecular Formula C2H5O3S
Molecular Weight (g/mol) 109.12
MDL Number MFCD00007529
InChI Key CCIVGXIOQKPBKL-UHFFFAOYSA-M
Synonym ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
PubChem CID 11668
ChEBI CHEBI:42465
SMILES CCS([O-])(=O)=O

Specifications

Melting Point -17.0°C
Color Yellow to Brown
Density 1.3430g/mL
Boiling Point 136°C to 140°C (3.0 hPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4325 to 1.4355
Linear Formula C2H5SO3H
Quantity 5 g
Beilstein 04, 5
Specific Gravity 1.343
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
IUPAC Name ethanesulfonate
Formula Weight 110.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethanesulfonic acid, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 209-843-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.