Alfa Aesar™ Ethanesulfonic acid, 96%

Manufacturer: Alfa Aesar™ 1489109

Quantity 10g

 View more versions of this product

Catalog No. AA1489109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 594-45-6
    Formula Weight 110.13
    MDL Number MFCD00007529
    UN Number UN2586
    Beilstein 1743071
    Synonym Ethylsulfonic acid
    Chemical Name or Material Ethanesulfonic acid

    Specifications

    Formula Weight 110.13
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 1743071
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 1 Danger
    Melting Point −17°C
    Quantity 10g
    Flash Point >110° (230°F)
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, LIQUID
    CAS 594-45-6
    UN Number UN2586
    Refractive Index 1.4350
    Density 1.343g/mL
    Boiling Point 122° to 123°C (0.01mmHg)
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00007529
    Synonym Ethylsulfonic acid
    Color Colorless to yellow
    Chemical Name or Material Ethanesulfonic acid
    Assay 96%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit