Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanesulfonic acid sodium salt, 98%, may cont. ca 2% water

Catalog No. p-7026047
Click to view available options
Quantity:
5 g
25 g

CAS: 5324-47-0 | C2H5NaO3S | 132.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5324-47-0
Molecular Formula C2H5NaO3S
Molecular Weight (g/mol) 132.109
MDL Number MFCD00066498
InChI Key KQFAFFYKLIBKDE-UHFFFAOYSA-M
Synonym sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
PubChem CID 4192170
IUPAC Name sodium;ethanesulfonate
SMILES CCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point ∼270°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 132.11
Percent Purity 98%
Assay may cont. ≈2% water
Chemical Name or Material Ethanesulfonic acid sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-194-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.