Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine, 98+%

Catalog No. p-7081646
Click to view available options
Quantity:
250 g
500 g
2500 g

Catalog No. AAA1169730

Supplier: Thermo Scientific Chemicals A1169730

Only null left
Add to Cart

CAS: 141-43-5 | C2H7NO | 61.08 g/mol

Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

Solubility
Miscible with water.

Notes
Store it away from heat, spark, open flames, hot surfaces and smoke. The storage area should be safe and enclosed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
SMILES NCCO

Specifications

Melting Point 10°C
Density 1.012
Boiling Point 169°C to 170°C
Flash Point 93°C (199°F)
Odor Ammonia-like
Refractive Index 1.454
Linear Formula HOCH2CH2NH2
Quantity 250 g
UN Number UN2491
Beilstein 505944
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,3727
Solubility Information Miscible with water.
IUPAC Name 2-aminoethan-1-ol
Formula Weight 61.08
Percent Purity ≥98%
Chemical Name or Material Ethanolamine
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ETHANOLAMINE
EINECSNumber 205-483-3
RTECSNumber KJ5775000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.