Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETHE1 Antibody (001), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB038699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038699 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038699 Supplier Novus Biologicals Supplier No. NBP290300G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ETHE1 Monoclonal specifically detects ETHE1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ETHE1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ETHE1 (O95571; Leu13-Ala254).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23474
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.