Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethenoadenosine Antibody (1G4), PE, Novus Biologicals™
SDP

Catalog No. NB600442PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600442PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600442PE Supplier Novus Biologicals Supplier No. NB600442PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ethenoadenosine Monoclonal specifically detects Ethenoadenosine in All Species samples. It is validated for Flow Cytometry.

Specifications

Antigen Ethenoadenosine
Applications Flow Cytometry
Classification Monoclonal
Clone 1G4
Conjugate PE
Dilution Flow Cytometry
Gene Alias C12H13N5O4, N6-Ethenoadenosine
Host Species Mouse
Immunogen Ethenoadenosine
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cell Biology
Primary or Secondary Primary
Test Specificity This is specific for ethenoadenosine and ethenodadenosine.
Target Species All species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.