Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethenocytidine Antibody (6F5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013801 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013801 Supplier Novus Biologicals Supplier No. NBP280728
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ethenocytidine Monoclonal specifically detects Ethenocytidine in All Species samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Ethenocytidine
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 6F5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS
Gene Alias C11H13N3O5
Host Species Mouse
Immunogen Ethenocytidine
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity All species.
Target Species All species
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.